Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kpna2
Entrez Gene ID
85245
Gene Name
karyopherin alpha 2
Gene Aliases
importin
Chromosome Location
Chr.10: 95200029 - 95212111 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270802.1 NP_001257731.1 3 - 4 347 77
NM_053483.2 NP_445935.2 3 - 4 322 77
GenBank mRNA AJ130946.1 - 2 - 3 226 77
AY779026.1 - 2 - 3 216 77
BC062026.1 - 3 - 4 322 77
BC089787.1 - 3 - 4 273 77
FM083931.1 - 3 - 4 322 77
FM115539.1 - 3 - 4 344 77

Gene Symbol
LOC100359600
Entrez Gene ID
100359600
Gene Name
karyopherin alpha 2-like
Gene Aliases
-
Chromosome Location
Chr.6: 10642705 - 10644693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729596.4 XP_002729642.1 1 - 1 342 77

View Full Product Details