Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc31a2
Entrez Gene ID
298091
Gene Name
solute carrier family 31 (copper transporter), member 2
Gene Aliases
-
Chromosome Location
Chr.5: 78120571 - 78131114 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033693.2 NP_001028865.2 2 - 3 190 82
GenBank mRNA BC099751.1 - 2 - 3 163 82

View Full Product Details