Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc30a3
Entrez Gene ID
366568
Gene Name
solute carrier family 30 (zinc transporter), member 3
Gene Aliases
ZnT3
Chromosome Location
Chr.6: 26629754 - 26650106 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013243.1 NP_001013261.1 2 - 3 357 65
XM_008764525.1 XP_008762747.1 3 - 4 357 65
XM_008764526.1 XP_008762748.1 3 - 4 402 65
XM_008764527.1 XP_008762749.1 3 - 4 331 65
XM_008764528.1 XP_008762750.1 3 - 4 437 65
XM_008764529.1 XP_008762751.1 4 - 5 532 65
GenBank mRNA AJ458940.1 - 1 - 2 180 65
AY538655.1 - 2 - 3 357 65
BC086513.1 - 3 - 4 329 65

View Full Product Details