The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002289.1 | NP_001002289.1 | 8 - 9 | 1413 | 80 |
XM_008764825.1 | XP_008763047.1 | 11 - 12 | 1921 | 80 | |
XM_008764826.1 | XP_008763048.1 | 11 - 12 | 2841 | 80 | |
GenBank mRNA | AJ781406.1 | - | 8 - 9 | 1413 | 80 |
FN668936.1 | - | 11 - 12 | 1921 | 80 | |
FN668937.1 | - | 11 - 12 | 1950 | 80 | |
FN668938.1 | - | 8 - 9 | 1635 | 80 |