Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Seli
Entrez Gene ID
362713
Gene Name
selenoprotein I
Gene Aliases
Ept1, RGD1560938
Chromosome Location
Chr.6: 27473748 - 27512287 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134754.1 NP_001128226.2 6 - 7 780 146
GenBank mRNA BC168235.1 - 6 - 7 780 146

View Full Product Details