Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Preb
Entrez Gene ID
58842
Gene Name
prolactin regulatory element binding
Gene Aliases
-
Chromosome Location
Chr.6: 26784088 - 26787875 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170708.1 NP_001164179.1 5 - 6 921 77
GenBank mRNA AF061817.1 - 5 - 6 865 77
BC078936.1 - 5 - 6 921 77

View Full Product Details