Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
29391
Gene Name
testis specific X-linked gene
Gene Aliases
-
Chromosome Location
Chr.X: 74269513 - 74283819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019203.1 NP_062076.1 5 - 6 420 111
XM_006257142.2 XP_006257204.1 6 - 7 500 111
GenBank mRNA X99797.1 - 5 - 6 420 111

View Full Product Details