The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017195.3 | NP_058891.1 | 2 - 3 | 810 | 79 |
GenBank mRNA | BC127502.1 | - | 2 - 3 | 658 | 79 |
FQ214107.1 | - | 2 - 3 | 645 | 79 | |
M16228.1 | - | 2 - 3 | 663 | 79 | |
M16736.1 | - | 2 - 3 | 646 | 79 | |
X06338.1 | - | 2 - 3 | 810 | 79 |