Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfi2
Entrez Gene ID
288038
Gene Name
antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5
Gene Aliases
-
Chromosome Location
Chr.11: 72135311 - 72158109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105872.1 NP_001099342.1 1 - 2 163 73

View Full Product Details