Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox3
Entrez Gene ID
100190895
Gene Name
reproductive homeobox on X chromosome 3
Gene Aliases
-
Chromosome Location
Chr.X: 123862565 - 123868309 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135607.1 NP_001129079.1 - 766 65
XM_008773382.1 XP_008771604.1 - 567 65
XM_008773383.1 XP_008771605.1 3 - 4 513 65
XM_008773384.1 XP_008771606.1 - 452 65
GenBank mRNA DQ058651.1 - - 766 65

Gene Symbol
LOC103690001
Entrez Gene ID
103690001
Gene Name
rhox homeobox family member 1-like
Gene Aliases
-
Chromosome Location
Chr.X: 124571933 - 124577677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257477.2 XP_006257539.1 3 - 4 513 65
XM_006257478.2 XP_006257540.1 2 - 3 452 65
XM_008773510.1 XP_008771732.1 3 - 4 567 65

View Full Product Details