The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212525.1 | NP_997690.1 | 4 - 5 | 292 | 65 |
XM_008759166.1 | XP_008757388.1 | 4 - 5 | 352 | 65 | |
GenBank mRNA | AY247021.1 | - | 4 - 5 | 292 | 65 |
FQ211365.1 | - | 4 - 5 | 298 | 65 |