Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhpn2
Entrez Gene ID
308516
Gene Name
rhophilin, Rho GTPase binding protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 91596350 - 91657395 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107505.1 NP_001100975.1 4 - 5 414 104
XM_006228887.2 XP_006228949.1 4 - 5 461 104
XM_008759160.1 XP_008757382.1 4 - 5 461 104

View Full Product Details