Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgals4
Entrez Gene ID
25474
Gene Name
lectin, galactoside-binding, soluble, 4
Gene Aliases
L-36, L36LBI
Chromosome Location
Chr.1: 87019808 - 87027720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012975.1 NP_037107.1 8 - 9 679 81
XM_008759113.1 XP_008757335.1 8 - 9 846 81
GenBank mRNA M73553.1 - 8 - 9 679 81

View Full Product Details