Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fv1
Entrez Gene ID
308568
Gene Name
Friend virus susceptibility 1
Gene Aliases
RGD1309326
Chromosome Location
Chr.1: 100220789 - 100226728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014018.2 NP_001014040.2 1 - 2 33 115
XM_008759362.1 XP_008757584.1 1 - 2 83 181
GenBank mRNA BC060592.1 - 1 - 2 37 115

View Full Product Details