Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St8sia2
Entrez Gene ID
117523
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
Siat8b
Chromosome Location
Chr.1: 135336374 - 135406699 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057156.1 NP_476497.1 2 - 3 155 113
GenBank mRNA L13445.1 - 2 - 3 155 113

View Full Product Details