Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rlbp1
Entrez Gene ID
293049
Gene Name
retinaldehyde binding protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 141097794 - 141111375 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106274.1 NP_001099744.1 3 - 4 579 64
XM_006229396.2 XP_006229458.1 3 - 4 495 64

View Full Product Details