Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpm3
Entrez Gene ID
309407
Gene Name
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases
-
Chromosome Location
Chr.1: 239741572 - 240524964 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191562.1 NP_001178491.1 - 3021 72
GenBank mRNA EF673688.1 - 1 - 2 188 72

View Full Product Details