Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep55
Entrez Gene ID
294074
Gene Name
centrosomal protein 55
Gene Aliases
RGD1305340
Chromosome Location
Chr.1: 256745250 - 256760794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025646.1 NP_001020817.1 4 - 5 672 103
XM_006231318.2 XP_006231380.1 4 - 5 652 103
XM_006231319.2 XP_006231381.1 4 - 5 766 103
XM_006231320.2 XP_006231382.1 4 - 5 609 103
GenBank mRNA BC098013.1 - 4 - 5 672 103

View Full Product Details