The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108480.1 | NP_001101950.1 | 4 - 5 | 524 | 59 |
XM_006229097.2 | XP_006229159.1 | 3 - 4 | 497 | 59 |