Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gltscr2
Entrez Gene ID
292624
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
-
Chromosome Location
Chr.1: 77836522 - 77843991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207591.1 NP_997474.1 7 - 8 871 57
GenBank mRNA AY535002.1 - 7 - 8 871 57
BC086359.1 - 7 - 8 875 57
FQ212888.1 - 7 - 8 906 57
FQ213201.1 - 7 - 8 880 57
FQ215499.1 - 5 - 6 572 57
FQ222789.1 - 7 - 8 881 57

View Full Product Details