Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gltscr2
Entrez Gene ID
292624
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
-
Chromosome Location
Chr.1: 77836522 - 77843991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207591.1 NP_997474.1 8 - 9 1068 62
GenBank mRNA AY535002.1 - 8 - 9 1068 62
BC086359.1 - 8 - 9 1072 62
FQ212888.1 - 8 - 9 1103 62
FQ213201.1 - 8 - 9 1077 62
FQ215499.1 - 6 - 7 769 62
FQ218937.1 - 8 - 9 1075 62
FQ219058.1 - 8 - 9 1075 62
FQ222789.1 - 8 - 9 1078 62

View Full Product Details