Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp6
Entrez Gene ID
363931
Gene Name
GTP binding protein 6 (putative)
Gene Aliases
RGD1305954
Chromosome Location
Chr.12: 52637000 - 52640869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135840.1 NP_001129312.1 4 - 5 650 78

Gene Symbol
LOC100912590
Entrez Gene ID
100912590
Gene Name
putative GTP-binding protein 6-like
Gene Aliases
-
Chromosome Location
Chr.12: 52670818 - 52674768 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751210.3 XP_003751258.2 - 730 78

View Full Product Details