Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wbscr27
Entrez Gene ID
688407
Gene Name
Williams Beuren syndrome chromosome region 27
Gene Aliases
-
Chromosome Location
Chr.12: 24766986 - 24776291 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109499.2 NP_001102969.1 3 - 4 388 86
XM_006249190.2 XP_006249252.1 4 - 5 474 86
XM_006249191.2 XP_006249253.1 4 - 5 474 86

View Full Product Details