Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crb1
Entrez Gene ID
304825
Gene Name
crumbs family member 1, photoreceptor morphogenesis associated
Gene Aliases
-
Chromosome Location
Chr.13: 56270781 - 56462834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107182.1 NP_001100652.1 3 - 4 967 109

View Full Product Details