The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012149.1 | NP_001012149.1 | 2 - 3 | 222 | 101 |
XM_006249960.2 | XP_006250022.1 | 2 - 3 | 257 | 101 | |
XM_006249961.2 | XP_006250023.1 | 2 - 3 | 257 | 101 | |
XM_006249962.2 | XP_006250024.1 | 2 - 3 | 258 | 101 | |
XM_006249963.2 | XP_006250025.1 | 2 - 3 | 259 | 101 | |
GenBank mRNA | BC088841.1 | - | 2 - 3 | 222 | 101 |
FQ220301.1 | - | 2 - 3 | 228 | 101 |