Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc45a3
Entrez Gene ID
304785
Gene Name
solute carrier family 45, member 3
Gene Aliases
RGD1309764
Chromosome Location
Chr.13: 48745860 - 48766283 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135868.1 NP_001129340.1 2 - 3 505 99
XM_006249765.2 XP_006249827.1 2 - 3 480 99
XM_006249766.2 XP_006249828.1 2 - 3 605 99
XM_006249767.2 XP_006249829.1 2 - 3 1083 99
XM_006249769.2 XP_006249831.1 2 - 3 1255 99

View Full Product Details