Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
24837
Gene Name
troponin T type 2 (cardiac)
Gene Aliases
CTTG, Ctt, RATCTTG, Tnnt3
Chromosome Location
Chr.13: 52662973 - 52680992 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012676.1 NP_036808.1 9 - 10 494 145
XM_006249822.1 XP_006249884.1 11 - 12 603 145
XM_006249823.1 XP_006249885.1 11 - 12 604 145
XM_006249825.2 XP_006249887.1 11 - 12 617 145
XM_006249828.1 XP_006249890.1 10 - 11 591 145
XM_006249829.1 XP_006249891.1 10 - 11 592 145
XM_006249833.2 XP_006249895.1 10 - 11 599 145
XM_006249835.1 XP_006249897.1 9 - 10 561 145
XM_006249836.1 XP_006249898.1 9 - 10 562 145
XM_006249838.1 XP_006249900.1 9 - 10 562 145
XM_006249840.2 XP_006249902.1 12 - 13 633 145
XM_006249842.2 XP_006249904.1 10 - 11 590 145
XM_008769535.1 XP_008767757.1 11 - 12 604 145
XM_008769536.1 XP_008767758.1 11 - 12 604 145
XM_008769537.1 XP_008767759.1 10 - 11 592 145
XM_008769538.1 XP_008767760.1 9 - 10 562 145
GenBank mRNA BC161855.1 - 10 - 11 599 145
EU295527.1 - 8 - 9 464 145
M26051.1 - 9 - 10 494 145
M26052.1 - 9 - 10 514 145

View Full Product Details