Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccndbp1
Entrez Gene ID
362201
Gene Name
cyclin D-type binding-protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 112950970 - 112961620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013204.1 NP_001013222.1 9 - 10 919 103
GenBank mRNA BC091256.1 - 9 - 10 919 103

View Full Product Details