Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35c1
Entrez Gene ID
311204
Gene Name
solute carrier family 35 (GDP-fucose transporter), member C1
Gene Aliases
-
Chromosome Location
Chr.3: 81361080 - 81369010 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107748.1 NP_001101218.1 1 - 2 1029 93
XM_006234568.2 XP_006234630.1 2 - 3 1238 93
XM_008762026.1 XP_008760248.1 2 - 3 1254 93

View Full Product Details