Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfyve19
Entrez Gene ID
499871
Gene Name
zinc finger, FYVE domain containing 19
Gene Aliases
-
Chromosome Location
Chr.3: 111013856 - 111021846 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034948.1 NP_001030120.1 5 - 6 582 60
XM_006234786.2 XP_006234848.1 5 - 6 518 60
XM_006234787.2 XP_006234849.1 5 - 6 518 60
XM_008762106.1 XP_008760328.1 5 - 6 582 60
GenBank mRNA BC099795.1 - 5 - 6 582 60

View Full Product Details