Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp19
Entrez Gene ID
311151
Gene Name
dual specificity phosphatase 19
Gene Aliases
-
Chromosome Location
Chr.3: 67849966 - 67862475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107739.2 NP_001101209.2 3 - 4 586 99

View Full Product Details