Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nop56
Entrez Gene ID
362214
Gene Name
NOP56 ribonucleoprotein
Gene Aliases
Nol5a
Chromosome Location
Chr.3: 122803728 - 122808564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025732.1 NP_001020903.1 3 - 4 263 62
GenBank mRNA BC099149.1 - 3 - 4 263 62

View Full Product Details