Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nop56
Entrez Gene ID
362214
Gene Name
NOP56 ribonucleoprotein
Gene Aliases
Nol5a
Chromosome Location
Chr.3: 122803728 - 122808564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025732.1 NP_001020903.1 10 - 11 1341 96
GenBank mRNA BC086568.1 - 7 - 8 1019 96
BC099149.1 - 10 - 11 1341 96
FQ224806.1 - 2 - 3 594 96
FQ232640.1 - 5 - 6 587 96

View Full Product Details