Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc67
Entrez Gene ID
315438
Gene Name
coiled-coil domain containing 67
Gene Aliases
RGD1306632
Chromosome Location
Chr.8: 14094742 - 14157235 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014087.1 NP_001014109.2 4 - 5 427 145
XM_008765933.1 XP_008764155.1 4 - 5 610 145
XM_008765934.1 XP_008764156.1 4 - 5 543 145
XM_008765935.1 XP_008764157.1 3 - 4 652 145
XM_008765936.1 XP_008764158.1 4 - 5 610 145
XM_008765937.1 XP_008764159.1 4 - 5 608 145
XM_008765938.1 XP_008764160.1 4 - 5 608 145
GenBank mRNA BC085333.1 - 4 - 5 427 145

View Full Product Details