The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013939.1 | NP_001013961.1 | 9 - 10 | 1811 | 134 |
GenBank mRNA | BC083865.1 | - | 9 - 10 | 1811 | 134 |