Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm17
Entrez Gene ID
291295
Gene Name
RNA binding motif protein 17
Gene Aliases
-
Chromosome Location
Chr.17: 70586343 - 70603289 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013058.1 NP_001013076.1 4 - 5 532 70
XM_006254205.2 XP_006254267.1 4 - 5 639 70
GenBank mRNA BC079248.1 - 4 - 5 532 70
FQ230702.1 - 3 - 4 391 70

View Full Product Details