Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2h2
Entrez Gene ID
294693
Gene Name
general transcription factor IIH, polypeptide 2
Gene Aliases
BTF2 p44
Chromosome Location
Chr.2: 30494792 - 30522717 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077428.1 NP_001070896.1 15 - 16 1127 154
XM_006231844.2 XP_006231906.1 15 - 16 1209 154
XM_006231845.2 XP_006231907.1 15 - 16 1367 154
GenBank mRNA BC126097.1 - 15 - 16 1127 154

View Full Product Details