Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcgr2b
Entrez Gene ID
289211
Gene Name
Fc fragment of IgG, low affinity IIb, receptor
Gene Aliases
CD32, FCRII, FcgammaRIIB2, Fcgr2
Chromosome Location
Chr.13: 89329298 - 89344169 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006250233.1 XP_006250295.1 5 - 6 837 103
XM_006250234.2 XP_006250296.1 6 - 7 850 103
GenBank mRNA EF424788.1 - 5 - 6 789 103

View Full Product Details