Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx59
Entrez Gene ID
289402
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 59
Gene Aliases
-
Chromosome Location
Chr.13: 53299872 - 53324824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190820.1 NP_001177749.1 4 - 5 1202 65
GenBank mRNA CO398251.1 - 3 - 4 550 65

View Full Product Details