Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Avpr1b
- Entrez Gene ID
-
29462
- Gene Name
- arginine vasopressin receptor 1B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 48367307 - 48379134 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017205.3 |
NP_058901.3 |
2 - 3 |
1576 |
60 |
GenBank mRNA |
D45400.1 |
- |
1 - 2 |
1105 |
60 |
U27322.1 |
- |
1 - 2 |
1469 |
60 |
- Gene Symbol
- LOC100909648
- Entrez Gene ID
-
100909648
- Gene Name
- vasopressin V1b receptor-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 48390434 - 48400768 on Build Rnor_6.0
View Full Product Details