The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134372.1 | NP_599199.1 | 8 - 9 | 942 | 79 |
GenBank mRNA | AB069781.1 | - | 8 - 9 | 942 | 79 |