The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024361.1 | NP_077337.1 | 14 - 15 | 2977 | 107 |
XM_008772130.1 | XP_008770352.1 | 10 - 11 | 1365 | 107 | |
GenBank mRNA | M92042.1 | - | 14 - 15 | 2977 | 107 |