Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mc2r
Entrez Gene ID
282839
Gene Name
melanocortin 2 receptor (adrenocorticotropic hormone)
Gene Aliases
-
Chromosome Location
Chr.18: 64166642 - 64178728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100491.1 NP_001093961.1 1 - 2 122 121

View Full Product Details