Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmo1
Entrez Gene ID
690253
Gene Name
slowmo homolog 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.18: 63202710 - 63217764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109570.1 NP_001103040.1 2 - 3 242 114
XM_006254884.2 XP_006254946.1 2 - 3 595 114
XM_006254885.2 XP_006254947.1 2 - 3 594 114
XM_006254886.2 XP_006254948.1 2 - 3 595 114

View Full Product Details