Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmo1
Entrez Gene ID
690253
Gene Name
slowmo homolog 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.18: 63202710 - 63217764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109570.1 NP_001103040.1 6 - 7 617 97

View Full Product Details