The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191836.1 | NP_001178765.1 | 5 - 6 | 866 | 59 |
GenBank mRNA | S80376.1 | - | 5 - 6 | 1268 | 59 |