Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh3tc2
Entrez Gene ID
307393
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
RGD1309038
Chromosome Location
Chr.18: 57286264 - 57403926 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254817.2 XP_006254879.1 10 - 11 1348 81
XM_006254818.2 XP_006254880.1 10 - 11 1227 81
XM_008772199.1 XP_008770421.1 10 - 11 1347 81
XM_008772200.1 XP_008770422.1 10 - 11 1347 81
XM_008772201.1 XP_008770423.1 10 - 11 1347 81
XM_008772202.1 XP_008770424.1 7 - 8 941 81
XM_225887.8 XP_225887.5 10 - 11 1347 81

View Full Product Details