Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap3s1
Entrez Gene ID
302290
Gene Name
adaptor-related protein complex 3, sigma 1 subunit
Gene Aliases
-
Chromosome Location
Chr.18: 40732445 - 40798059 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106933.1 NP_001100403.1 5 - 6 549 109

View Full Product Details