Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a18
Entrez Gene ID
681896
Gene Name
solute carrier family 25 (glutamate carrier), member 18
Gene Aliases
-
Chromosome Location
Chr.4: 153330069 - 153350447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044280.2 NP_001037745.2 9 - 10 1092 116
GenBank mRNA BC094517.1 - 9 - 10 1037 116

View Full Product Details