Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Commd6
Entrez Gene ID
498559
Gene Name
COMM domain containing 6
Gene Aliases
RGD1560745
Chromosome Location
Chr.15: 86136096 - 86142672 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770892.1 XP_008769114.1 1 - 2 188 75

View Full Product Details